Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124561.3 | NP_199995.1 | 15 - 16 | 1622 | 93 | |
GenBank mRNA | AF216580.1 | - | 15 - 16 | 1337 | 93 | |
AY099708.1 | - | 15 - 16 | 1293 | 93 | ||
AY128901.1 | - | 15 - 16 | 1228 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |