Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344986.1 | NP_001318786.1 | 2 - 3 | 391 | 116 | |
NM_001344987.1 | NP_001332093.1 | 2 - 3 | 413 | 116 | ||
NM_124596.2 | NP_200030.1 | 1 - 2 | 214 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |