Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344986.1 | NP_001318786.1 | 8 - 9 | 1848 | 73 | |
NM_001344987.1 | NP_001332093.1 | 8 - 9 | 1885 | 73 | ||
NM_124596.2 | NP_200030.1 | 7 - 8 | 1671 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |