Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203591.2 | NP_001190520.1 | 4 - 5 | 672 | 71 | |
NM_001344990.1 | NP_001331264.1 | 4 - 5 | 559 | 71 | ||
NM_124602.4 | NP_200036.1 | 3 - 4 | 775 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |