Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTI65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036984.1 | NP_001032061.1 | 3 - 4 | 930 | 112 | |
NM_124609.4 | NP_200043.2 | 3 - 4 | 968 | 112 | ||
GenBank mRNA | AY081282.1 | - | 3 - 4 | 932 | 112 | |
AY128731.1 | - | 3 - 4 | 784 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |