Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO70A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124613.4 | NP_200047.3 | 7 - 8 | 1407 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |