Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344997.1 | NP_001331016.1 | 1 - 2 | 448 | 75 | |
NM_124625.2 | NP_200059.2 | 2 - 3 | 652 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |