Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124692.3 | NP_200125.1 | 7 - 8 | 1843 | 74 | |
GenBank mRNA | AF370139.1 | - | 7 - 8 | 1843 | 74 | |
BT000991.1 | - | 6 - 7 | 1450 | 74 | ||
Y16327.1 | - | 7 - 8 | 1754 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |