Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124699.3 | NP_200132.2 | 1 - 2 | 358 | 123 | |
GenBank mRNA | AY161286.1 | - | 1 - 2 | 198 | 123 | |
AY519523.1 | - | 1 - 2 | 59 | 123 | ||
BT024672.1 | - | 1 - 2 | 59 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |