Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345044.1 | NP_001331493.1 | 1 - 2 | 713 | 69 | |
NM_124700.3 | NP_200133.2 | 1 - 2 | 713 | 69 | ||
GenBank mRNA | AJ630498.1 | - | 1 - 2 | 399 | 69 | |
AY568670.1 | - | 1 - 2 | 399 | 69 | ||
DQ868373.1 | - | 1 - 2 | 399 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |