Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203600.1 | NP_001190529.1 | 19 - 20 | 6683 | 62 | |
NM_001203601.1 | NP_001190530.1 | 19 - 20 | 6683 | 62 | ||
NM_124725.5 | NP_200158.2 | 19 - 20 | 6698 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |