Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124732.2 | NP_200164.1 | 2 - 3 | 659 | 117 | |
GenBank mRNA | DQ447070.1 | - | 2 - 3 | 575 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |