Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSFA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124786.4 | NP_200218.1 | 1 - 2 | 419 | 121 | |
GenBank mRNA | AY065226.1 | - | 1 - 2 | 418 | 121 | |
AY096552.1 | - | 1 - 2 | 384 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |