Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336567.1 | NP_001325149.1 | 50 - 51 | 5989 | 81 | |
NM_129117.5 | NP_565811.2 | 50 - 51 | 5683 | 81 | ||
GenBank mRNA | AF367246.1 | - | 50 - 51 | 5429 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |