Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA07 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336568.1 | NP_001325223.1 | 2 - 3 | 677 | 86 | |
NM_001336569.1 | NP_001325221.1 | 2 - 3 | 538 | 86 | ||
NM_001336570.1 | NP_001325222.1 | 1 - 2 | 376 | 86 | ||
NM_129120.4 | NP_565813.1 | 2 - 3 | 556 | 86 | ||
GenBank mRNA | AJ488284.1 | - | 2 - 3 | 538 | 86 | |
AY059724.1 | - | 2 - 3 | 527 | 86 | ||
AY133807.1 | - | 2 - 3 | 401 | 86 | ||
KJ138872.1 | - | 2 - 3 | 401 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |