Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129124.4 | NP_181112.1 | 1 - 2 | 374 | 89 | |
GenBank mRNA | AY074357.1 | - | 1 - 2 | 346 | 89 | |
AY096691.1 | - | 1 - 2 | 138 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |