Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129132.2 | NP_181117.1 | 2 - 3 | 1481 | 101 | |
GenBank mRNA | AJ973177.1 | - | 2 - 3 | 1269 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |