Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129135.3 | NP_181120.2 | 3 - 4 | 621 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |