Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129136.5 | NP_181121.1 | 2 - 3 | 650 | 111 | |
GenBank mRNA | AY088044.1 | - | 2 - 3 | 470 | 111 | |
BT029178.1 | - | 2 - 3 | 456 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |