Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129188.4 | NP_565843.1 | 2 - 3 | 861 | 68 | |
GenBank mRNA | AY054182.1 | - | 2 - 3 | 312 | 68 | |
AY066040.1 | - | 2 - 3 | 218 | 68 | ||
AY086551.1 | - | 2 - 3 | 341 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |