Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129188.4 | NP_565843.1 | 6 - 7 | 1307 | 76 | |
GenBank mRNA | AY054182.1 | - | 6 - 7 | 758 | 76 | |
AY066040.1 | - | 6 - 7 | 664 | 76 | ||
AY086551.1 | - | 6 - 7 | 787 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |