Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129195.6 | NP_181179.2 | 13 - 14 | 3384 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |