Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129195.6 | NP_181179.2 | 15 - 16 | 3599 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |