Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129207.5 | NP_181190.3 | 2 - 3 | 423 | 95 | |
GenBank mRNA | AY286009.1 | - | 1 - 2 | 130 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |