Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECP63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336603.1 | NP_001324810.1 | 1 - 2 | 955 | 95 | |
NM_129219.3 | NP_181202.1 | 1 - 2 | 961 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |