Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336608.1 | NP_001324358.1 | 11 - 12 | 1473 | 105 | |
NM_129229.4 | NP_181212.2 | 12 - 13 | 1365 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |