Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129265.3 | NP_181246.1 | 2 - 3 | 1068 | 72 | |
GenBank mRNA | BT015044.1 | - | 2 - 3 | 953 | 72 | |
BT015838.1 | - | 2 - 3 | 922 | 72 | ||
KJ138863.1 | - | 2 - 3 | 922 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |