Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129310.3 | NP_181291.1 | 3 - 4 | 1516 | 102 | |
GenBank mRNA | AB017876.1 | - | 3 - 4 | 1518 | 102 | |
AF436828.1 | - | 3 - 4 | 1433 | 102 | ||
AY034983.1 | - | 3 - 4 | 1432 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |