Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084551.1 | NP_001078020.1 | 11 - 12 | 2022 | 113 | |
NM_129359.4 | NP_565879.1 | 11 - 12 | 2023 | 113 | ||
GenBank mRNA | AF359240.1 | - | 10 - 11 | 1930 | 113 | |
AY048248.1 | - | 11 - 12 | 1997 | 113 | ||
AY139799.1 | - | 10 - 11 | 1914 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |