Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129370.5 | NP_181349.1 | 1 - 2 | 258 | 106 | |
GenBank mRNA | AF236826.1 | - | 1 - 2 | 177 | 106 | |
AY052215.1 | - | 1 - 2 | 200 | 106 | ||
AY060491.1 | - | 1 - 2 | 158 | 106 | ||
AY088244.1 | - | 1 - 2 | 223 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |