Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129424.5 | NP_181401.1 | 3 - 4 | 1011 | 92 | |
GenBank mRNA | AY092984.1 | - | 3 - 4 | 851 | 92 | |
AY128807.1 | - | 3 - 4 | 744 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |