Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129433.5 | NP_181410.1 | 3 - 4 | 605 | 106 | |
GenBank mRNA | AK227695.1 | - | 3 - 4 | 532 | 106 | |
AY087221.1 | - | 3 - 4 | 535 | 106 | ||
BT006420.1 | - | 3 - 4 | 463 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |