Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202779.1 | NP_001189708.1 | 7 - 8 | 1158 | 83 | |
NM_129454.4 | NP_181430.2 | 7 - 8 | 1271 | 83 | ||
NM_201908.1 | NP_973637.1 | 7 - 8 | 1102 | 83 | ||
GenBank mRNA | AJ551466.1 | - | 7 - 8 | 1102 | 83 | |
BT022038.1 | - | 7 - 8 | 1123 | 83 | ||
BT023745.1 | - | 7 - 8 | 1102 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |