Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATATH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129476.3 | NP_565900.1 | 3 - 4 | 824 | 65 | |
NM_129477.1 | NP_565901.1 | 3 - 4 | 824 | 65 | ||
GenBank mRNA | AY048299.1 | - | 3 - 4 | 824 | 65 | |
BT002715.1 | - | 3 - 4 | 652 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |