Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129478.5 | NP_565902.1 | 10 - 11 | 1090 | 144 | |
GenBank mRNA | AF369573.1 | - | 10 - 11 | 960 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |