Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129478.5 | NP_565902.1 | 13 - 14 | 1371 | 81 | |
GenBank mRNA | AF369573.1 | - | 13 - 14 | 1241 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |