Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PnsL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036437.3 | NP_001031514.2 | 1 - 2 | 179 | 72 | |
NM_001084553.1 | NP_001078022.1 | 1 - 2 | 179 | 72 | ||
NM_129505.4 | NP_565906.1 | 1 - 2 | 197 | 72 | ||
GenBank mRNA | AY039968.1 | - | 1 - 2 | 93 | 72 | |
AY085632.1 | - | 1 - 2 | 88 | 72 | ||
AY133798.1 | - | 1 - 2 | 60 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |