Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129506.2 | NP_181480.1 | 1 - 2 | 615 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Arabidopsis thaliana Probe spans exons |