Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGT-I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129513.4 | NP_181487.1 | 4 - 5 | 614 | 98 | |
GenBank mRNA | BT002465.1 | - | 4 - 5 | 562 | 98 | |
BT008464.1 | - | 4 - 5 | 519 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |