Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336807.1 | NP_001324597.1 | 10 - 11 | 1436 | 108 | |
NM_001336808.1 | NP_001324596.1 | 10 - 11 | 1436 | 108 | ||
NM_129551.4 | NP_181522.1 | 10 - 11 | 1608 | 108 | ||
GenBank mRNA | BT000443.1 | - | 10 - 11 | 1412 | 108 | |
BT010348.1 | - | 10 - 11 | 1381 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |