Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129551.4 | NP_181522.1 | 16 - 17 | 2193 | 143 | |
GenBank mRNA | BT000443.1 | - | 16 - 17 | 1997 | 143 | |
BT010348.1 | - | 16 - 17 | 1966 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |