Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336807.1 | NP_001324597.1 | 8 - 9 | 1239 | 67 | |
NM_001336808.1 | NP_001324596.1 | 8 - 9 | 1239 | 67 | ||
NM_129551.4 | NP_181522.1 | 8 - 9 | 1411 | 67 | ||
GenBank mRNA | BT000443.1 | - | 8 - 9 | 1215 | 67 | |
BT010348.1 | - | 8 - 9 | 1184 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |