Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129567.4 | NP_565923.1 | 4 - 5 | 743 | 69 | |
GenBank mRNA | AK175658.1 | - | 4 - 5 | 573 | 69 | |
AK176037.1 | - | 4 - 5 | 573 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |