Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129611.5 | NP_181581.1 | 4 - 5 | 696 | 78 | |
GenBank mRNA | AY142634.1 | - | 4 - 5 | 540 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |