Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129617.5 | NP_181587.2 | 5 - 6 | 873 | 75 | |
GenBank mRNA | BT022032.1 | - | 5 - 6 | 582 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |