Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129629.3 | NP_181599.1 | 3 - 4 | 912 | 71 | |
GenBank mRNA | AF305721.1 | - | 3 - 4 | 327 | 71 | |
AJ279079.1 | - | 3 - 4 | 327 | 71 | ||
BT043482.1 | - | 3 - 4 | 327 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |