Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336861.1 | NP_001324304.1 | 1 - 2 | 545 | 74 | |
NM_129636.3 | NP_181606.1 | 1 - 2 | 446 | 74 | ||
GenBank mRNA | AB493584.1 | - | 1 - 2 | 446 | 74 | |
DQ056575.1 | - | 1 - 2 | 446 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |