Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129647.4 | NP_181616.3 | 14 - 15 | 2042 | 117 | |
GenBank mRNA | AY081315.1 | - | 14 - 15 | 1833 | 117 | |
BT010364.1 | - | 13 - 14 | 1784 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |