Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129658.4 | NP_181626.1 | 3 - 4 | 1542 | 103 | |
GenBank mRNA | AY054532.1 | - | 3 - 4 | 1484 | 103 | |
BT000159.1 | - | 2 - 3 | 1270 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |