Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129716.4 | NP_181684.1 | 2 - 3 | 567 | 59 | |
GenBank mRNA | AF378875.1 | - | 2 - 3 | 287 | 59 | |
AF446861.1 | - | 2 - 3 | 265 | 59 | ||
AJ347732.1 | - | 2 - 3 | 265 | 59 | ||
AY044322.1 | - | 2 - 3 | 265 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |